Assay Details
Gene Symbol: TNFRSF14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284454.1 | NP_001271383.1 | 179 | 85 | ||
XM_005620424.3 | XP_005620481.1 | 2804 | 85 | |||
XM_005620425.3 | XP_005620482.1 | 2811 | 85 | |||
XM_005620426.3 | XP_005620483.1 | 2812 | 85 | |||
XM_022418017.1 | XP_022273725.1 | 988 | 85 | |||
GenBank mRNA | JX435818.1 | - | 179 | 85 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
