Assay Details
Gene Symbol: TVP23A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_547129.6 | XP_547129.2 | 1 - 2 | 152 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_547129.6 transcripts.
Set Membership: |
![]() ![]() ![]() |
