Assay Details
Gene Symbol: CPTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639055.4 | XP_003639103.1 | 80 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_003639055.4 transcripts.
Set Membership: |
![]() ![]() |
