Assay Details
Gene Symbol: BNIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022412057.1 | XP_022267765.1 | 4 - 5 | 414 | 57 | |
XM_844054.6 | XP_849147.4 | 4 - 5 | 525 | 57 | ||
GenBank mRNA | DQ926865.1 | - | 4 - 5 | 390 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
