Assay Details
Gene Symbol: GFOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639071.4 | XP_003639119.1 | 2 - 3 | 498 | 66 | |
XM_005620815.3 | XP_005620872.1 | 3 - 4 | 677 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
