Assay Details
Gene Symbol: RP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014111448.2 | XP_013966923.1 | 3 - 4 | 1079 | 66 | |
XM_548972.6 | XP_548972.2 | 3 - 4 | 1080 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
