Assay Details
Gene Symbol: MRPL55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005628353.3 | XP_005628410.1 | 2 - 3 | 192 | 75 | |
XM_005628354.3 | XP_005628411.1 | 2 - 3 | 169 | 75 | ||
XM_843940.5 | XP_849033.2 | 2 - 3 | 159 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005628353.3, XM_005628354.3, XM_843940.5 transcripts.
Set Membership: |
![]() ![]() ![]() |
