Assay Details
Gene Symbol: TRAPPC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014113950.2 | XP_013969425.1 | 257 | 68 | ||
XM_844929.5 | XP_850022.1 | 291 | 68 |

More Information
Set Membership: |
![]() ![]() |
