Assay Details
Gene Symbol: SPINK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_845464.5 | XP_850557.1 | 2 - 3 | 161 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
