Assay Details
Gene Symbol: PTGES2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001131050.1 | NP_001124522.1 | 4 - 5 | 768 | 60 | |
XM_005625033.3 | XP_005625090.1 | 4 - 5 | 609 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
