Assay Details
Gene Symbol: ARPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252380.1 | NP_001239309.1 | 2 - 3 | 230 | 71 | |
XM_005618244.1 | XP_005618301.1 | 2 - 3 | 541 | 71 | ||
XM_022415725.1 | XP_022271433.1 | 2 - 3 | 144 | 71 | ||
GenBank mRNA | DN434974.1 | - | 2 - 3 | 239 | 71 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022415725.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
