Assay Details
Gene Symbol: SPACA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005625181.2 | XP_005625238.1 | 5 - 6 | 881 | 73 | |
XM_005625186.2 | XP_005625243.1 | 4 - 5 | 585 | 73 | ||
XM_022423660.1 | XP_022279368.1 | 5 - 6 | 659 | 73 | ||
XM_022423661.1 | XP_022279369.1 | 5 - 6 | 858 | 73 | ||
XM_022423662.1 | XP_022279370.1 | 4 - 5 | 834 | 73 | ||
XM_022423663.1 | XP_022279371.1 | 3 - 4 | 914 | 73 | ||
XM_022423664.1 | XP_022279372.1 | 5 - 6 | 701 | 73 | ||
XM_022423665.1 | XP_022279373.1 | 3 - 4 | 703 | 73 | ||
XM_022423666.1 | XP_022279374.1 | 6 - 7 | 753 | 73 | ||
XM_022423667.1 | XP_022279375.1 | 3 - 4 | 636 | 73 | ||
XM_022423668.1 | XP_022279376.1 | 6 - 7 | 750 | 73 | ||
XM_022423669.1 | XP_022279377.1 | 6 - 7 | 977 | 73 | ||
XM_845436.5 | XP_850529.1 | 3 - 4 | 899 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
