Assay Details
Gene Symbol: LOC106557520 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014114467.2 | XP_013969942.1 | 4 - 5 | 897 | 97 | |
XM_022420019.1 | XP_022275727.1 | 4 - 5 | 901 | 97 | ||
XM_022420020.1 | XP_022275728.1 | 4 - 5 | 530 | 97 | ||
XM_022420021.1 | XP_022275729.1 | 4 - 5 | 908 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
