Assay Details
Gene Symbol: LOC106557520 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014114467.2 | XP_013969942.1 | 12 - 13 | 1536 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC106557520 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014114467.2 | XP_013969942.1 | 12 - 13 | 1536 | 83 |
Set Membership: |
![]() ![]() ![]() ![]() |