Assay Details
Gene Symbol: AKAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005641354.3 | XP_005641411.1 | 5 - 6 | 2546 | 61 | |
XM_846452.5 | XP_851545.1 | 5 - 6 | 2545 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
