Assay Details
Gene Symbol: ADA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022411277.1 | XP_022266985.1 | 1 - 2 | 119 | 57 | |
XM_022411278.1 | XP_022266986.1 | 1 - 2 | 119 | 57 | ||
XM_849704.5 | XP_854797.1 | 1 - 2 | 119 | 57 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022411277.1, XM_022411278.1, XM_849704.5 transcripts.
Set Membership: |
![]() ![]() ![]() |
