Assay Details
Gene Symbol: ADA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022411277.1 | XP_022266985.1 | 2 - 3 | 452 | 101 | |
XM_022411278.1 | XP_022266986.1 | 2 - 3 | 452 | 101 | ||
XM_849704.5 | XP_854797.1 | 2 - 3 | 452 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
