Assay Details
Gene Symbol: NCMAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022413250.1 | XP_022268958.1 | 2 - 3 | 293 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
