Assay Details
Gene Symbol: PLPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639749.3 | XP_003639797.1 | 2 - 3 | 271 | 91 | |
XM_022407336.1 | XP_022263044.1 | 2 - 3 | 273 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022407336.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
