Assay Details
Gene Symbol: PLPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003639749.3 | XP_003639797.1 | 5 - 6 | 790 | 93 | |
XM_022407336.1 | XP_022263044.1 | 5 - 6 | 792 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
