Assay Details
Gene Symbol: C26H12orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014107973.2 | XP_013963448.1 | 2 - 3 | 229 | 78 | |
XM_849238.4 | XP_854331.1 | 2 - 3 | 229 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
