Assay Details
Gene Symbol: CDC42EP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_849090.5 | XP_854183.1 | 2 - 3 | 417 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_849090.5 transcripts.
Set Membership: |
![]() ![]() ![]() |
