Assay Details
Gene Symbol: ASPHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022410513.1 | XP_022266221.1 | 4 - 5 | 1339 | 84 | |
XM_022410514.1 | XP_022266222.1 | 4 - 5 | 1821 | 84 | ||
XM_022410515.1 | XP_022266223.1 | 3 - 4 | 1275 | 84 | ||
XM_849500.5 | XP_854593.2 | 3 - 4 | 1785 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
