Assay Details
Gene Symbol: SELENOW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115012.1 | NP_001108484.1 | 5 - 6 | 311 | 57 | |
GenBank mRNA | CN005642.1 | - | 5 - 6 | 334 | 57 | |
DN370714.1 | - | 5 - 6 | 308 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
