Assay Details
Gene Symbol: PKIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003433669.4 | XP_003433717.1 | 3 - 4 | 421 | 84 | |
XM_005638060.3 | XP_005638117.2 | 2 - 3 | 191 | 84 | ||
XM_005638062.2 | XP_005638119.1 | 3 - 4 | 344 | 84 | ||
XM_022412474.1 | XP_022268182.1 | 2 - 3 | 287 | 84 | ||
XM_848290.4 | XP_853383.2 | 2 - 3 | 281 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
