Assay Details
Gene Symbol: NECTIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022425008.1 | XP_022280716.1 | 7 - 8 | 1253 | 61 | |
XM_847558.5 | XP_852651.1 | 7 - 8 | 1489 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
