Assay Details
Gene Symbol: ZNF653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_848511.5 | XP_853604.1 | 8 - 9 | 1704 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ZNF653 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_848511.5 | XP_853604.1 | 8 - 9 | 1704 | 144 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |