Assay Details
Gene Symbol: TEDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005621683.3 | XP_005621740.1 | 6 - 7 | 841 | 72 | |
XM_005621684.3 | XP_005621741.1 | 6 - 7 | 841 | 72 | ||
XM_014114689.2 | XP_013970164.1 | 5 - 6 | 692 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
