Assay Details
Gene Symbol: ATP5F1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005617159.2 | XP_005617216.1 | 1 | 322 | 103 | |
XM_005617160.2 | XP_005617217.1 | 1 | 322 | 103 | ||
XM_853139.4 | XP_858232.1 | 1 | 322 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
