Assay Details
Gene Symbol: SUV39H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005617110.3 | XP_005617167.1 | 1045 | 80 | ||
XM_005617111.2 | XP_005617168.1 | 1115 | 80 | |||
XM_005617112.3 | XP_005617169.1 | 1176 | 80 | |||
XM_535179.6 | XP_535179.2 | 1235 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
