Assay Details
Gene Symbol: MYL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022424049.1 | XP_022279757.1 | 1 | 526 | 77 | |
XM_851657.5 | XP_856750.2 | 1 | 591 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
