Assay Details
Gene Symbol: DIABLO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079767.1 | NP_001073235.1 | 533 | 109 | ||
XM_022410149.1 | XP_022265857.1 | 491 | 109 | |||
GenBank mRNA | EF088670.1 | - | 533 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
