Assay Details
Gene Symbol: EIF5A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022414133.1 | XP_022269841.1 | 1 - 2 | 216 | 92 | |
XM_022414134.1 | XP_022269842.1 | 1 - 2 | 202 | 92 | ||
XM_022414135.1 | XP_022269843.1 | 1 - 2 | 202 | 92 | ||
XM_022414136.1 | XP_022269844.1 | 1 - 2 | 202 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
