Assay Details
Gene Symbol: GABPB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014120613.2 | XP_013976088.1 | 3 - 4 | 1509 | 60 | |
XM_014120614.2 | XP_013976089.1 | 4 - 5 | 1709 | 60 | ||
XM_014120615.2 | XP_013976090.1 | 3 - 4 | 1530 | 60 | ||
XM_014120616.2 | XP_013976091.1 | 4 - 5 | 1663 | 60 | ||
XM_014120617.2 | XP_013976092.1 | 4 - 5 | 1631 | 60 | ||
XM_014120618.2 | XP_013976093.1 | 3 - 4 | 1477 | 60 | ||
XM_014120620.2 | XP_013976095.1 | 3 - 4 | 1509 | 60 | ||
XM_022404990.1 | XP_022260698.1 | 3 - 4 | 476 | 60 | ||
XM_022404991.1 | XP_022260699.1 | 4 - 5 | 1593 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
