Assay Details
Gene Symbol: TRIB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005634904.3 | XP_005634961.1 | 3 - 4 | 769 | 63 | |
XM_542943.5 | XP_542943.2 | 3 - 4 | 643 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
