Assay Details
Gene Symbol: CKMT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022413031.1 | XP_022268739.1 | 7 - 8 | 1166 | 78 | |
XM_535451.6 | XP_535451.1 | 6 - 7 | 1115 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
