Assay Details
Gene Symbol: LRRC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005625263.3 | XP_005625320.1 | 3 - 4 | 2390 | 91 | |
XM_005625265.3 | XP_005625322.1 | 4 - 5 | 2448 | 91 | ||
XM_022423747.1 | XP_022279455.1 | 2 - 3 | 2250 | 91 | ||
XM_548430.5 | XP_548430.2 | 2 - 3 | 2283 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
