Assay Details
Gene Symbol: LAMTOR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014114227.2 | XP_013969702.1 | 156 | 72 | ||
XM_022419665.1 | XP_022275373.1 | 156 | 72 | |||
XM_536864.6 | XP_536864.1 | 156 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014114227.2, XM_022419665.1, XM_536864.6 transcripts.
Set Membership: |
![]() ![]() |
