Assay Details
Gene Symbol: LAMTOR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014114227.2 | XP_013969702.1 | 233 | 67 | ||
XM_022419665.1 | XP_022275373.1 | 233 | 67 | |||
XM_536864.6 | XP_536864.1 | 233 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
