Assay Details
Gene Symbol: SRP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252291.1 | NP_001239220.1 | 1 | 364 | 108 | |
GenBank mRNA | DN398351.1 | - | 1 | 364 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
