Assay Details
Gene Symbol: DBI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197047.1 | NP_001183976.1 | 3 - 4 | 269 | 144 | |
XM_014121370.2 | XP_013976845.1 | 3 - 4 | 322 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
