Assay Details
Gene Symbol: NACA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252257.1 | NP_001239186.1 | 1 | 531 | 73 | |
NM_001252258.1 | NP_001239187.1 | 1 | 440 | 73 | ||
XM_022423989.1 | XP_022279697.1 | 1 | 446 | 73 | ||
XM_022423990.1 | XP_022279698.1 | 1 | 397 | 73 | ||
XM_022423991.1 | XP_022279699.1 | 577 | 73 | |||
XM_022423992.1 | XP_022279700.1 | 1 | 511 | 73 | ||
GenBank mRNA | CO584962.1 | - | 1 | 391 | 73 | |
DT536457.1 | - | 1 | 447 | 73 | ||
GR888023.1 | - | 1 | 508 | 73 | ||
GR892750.1 | - | 531 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
