Assay Details
Gene Symbol: LOC476047 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005631587.3 | XP_005631644.1 | 2 - 3 | 579 | 143 | |
XM_005631588.3 | XP_005631645.1 | 2 - 3 | 558 | 143 | ||
XM_022405770.1 | XP_022261478.1 | 1 - 2 | 676 | 143 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
