Assay Details
Gene Symbol: HS2ST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005621962.3 | XP_005622019.1 | 4 - 5 | 892 | 166 | |
XM_537087.6 | XP_537087.3 | 4 - 5 | 585 | 166 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
