Assay Details
Gene Symbol: MYCL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_539578.6 | XP_539578.2 | 1 - 2 | 102 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
