Assay Details
Gene Symbol: KCNJ15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005638910.3 | XP_005638967.1 | 4 | 1676 | 153 | |
XM_014109822.2 | XP_013965297.1 | 5 | 1099 | 153 | ||
XM_014109823.2 | XP_013965298.1 | 4 | 1683 | 153 | ||
XM_014109825.2 | XP_013965300.1 | 5 | 1092 | 153 | ||
XM_014109827.2 | XP_013965302.1 | 3 | 1628 | 153 | ||
XM_014109828.2 | XP_013965303.1 | 4 | 1102 | 153 | ||
XM_014109829.2 | XP_013965304.1 | 4 | 1046 | 153 | ||
XM_014109830.2 | XP_013965305.1 | 4 | 994 | 153 | ||
XM_014109831.2 | XP_013965306.1 | 4 | 1190 | 153 | ||
XM_022413188.1 | XP_022268896.1 | 5 | 1173 | 153 | ||
XM_022413189.1 | XP_022268897.1 | 5 | 1122 | 153 | ||
XM_022413190.1 | XP_022268898.1 | 4 | 1039 | 153 | ||
XM_022413191.1 | XP_022268899.1 | 4 | 978 | 153 | ||
XM_022413192.1 | XP_022268900.1 | 4 | 865 | 153 | ||
XM_022413193.1 | XP_022268901.1 | 3 | 997 | 153 | ||
XM_022413194.1 | XP_022268902.1 | 3 | 1140 | 153 | ||
XM_544884.6 | XP_544884.2 | 3 | 946 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
