Assay Details
Gene Symbol: BOLL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005640499.3 | XP_005640556.2 | 4 - 5 | 352 | 149 | |
XM_014111035.2 | XP_013966510.1 | 4 - 5 | 352 | 149 | ||
XM_022415179.1 | XP_022270887.1 | 4 - 5 | 352 | 149 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
