Assay Details
Gene Symbol: RASGEF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022411779.1 | XP_022267487.1 | 6 - 7 | 986 | 99 | |
XM_022411780.1 | XP_022267488.1 | 6 - 7 | 1415 | 99 | ||
XM_022411781.1 | XP_022267489.1 | 7 - 8 | 1203 | 99 | ||
XM_022411782.1 | XP_022267490.1 | 6 - 7 | 1064 | 99 | ||
XM_022411783.1 | XP_022267491.1 | 6 - 7 | 1225 | 99 | ||
XM_022411784.1 | XP_022267492.1 | 6 - 7 | 1041 | 99 | ||
XM_022411785.1 | XP_022267493.1 | 6 - 7 | 1093 | 99 | ||
XM_534955.6 | XP_534955.4 | 6 - 7 | 857 | 99 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
