Assay Details
Gene Symbol: PLPPR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022407313.1 | XP_022263021.1 | 6 | 1536 | 147 | |
XM_022407314.1 | XP_022263022.1 | 7 | 1451 | 147 | ||
XM_542214.7 | XP_542214.4 | 7 | 1373 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
