Assay Details
Gene Symbol: MITF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003337.1 | NP_001003337.1 | 272 | 69 | ||
XM_005632016.2 | XP_005632073.1 | 487 | 69 | |||
XM_005632018.2 | XP_005632075.1 | 487 | 69 | |||
XM_005632019.3 | XP_005632076.1 | 155 | 69 | |||
GenBank mRNA | AY240952.1 | - | 272 | 69 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() |
